missing translation for 'onlineSavingsMsg'
Learn More

Zinc nitride, 99% (metals basis)

Product Code. 11329219
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329219 2 g 2g
11319219 10 g 10g
15412987 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11329219 Supplier Thermo Scientific Alfa Aesar Supplier No. 088212.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1313-49-1 | N2Zn3 | 224.15 g/mol

Zinc nitride is useful in optical and electronic devices. It is doped with oxygen to prepare a potential nitride-based transparent conductor, which is used in semiconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-49-1
Molecular Formula N2Zn3
Molecular Weight (g/mol) 224.15
MDL Number MFCD00061327
InChI Key AKJVMGQSGCSQBU-UHFFFAOYSA-N
Synonym zinc nitride, zinc nitride zn3n2
PubChem CID 12130759
SMILES [Zn++].[N-]=[Zn].[N-]=[Zn]

Specifications

Physical Form Powder
Quantity 2 g
Solubility Information Insoluble in water. Soluble in hydrochloric acid.
IUPAC Name zinc;azanidylidenezinc
Formula Weight 224.12
Percent Purity 99%
Density 6.22 g/mL
Assay (metals basis)
Chemical Name or Material Zinc nitride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.