missing translation for 'onlineSavingsMsg'
Learn More

Zinc Nitrate Hexahydrate, Extra Pure, SLR, Fisher Chemical™

Product Code. 10796301
Click to view available options
Quantity:
250 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 10196-18-6
Molecular Formula H12N2O12Zn
Molecular Weight (g/mol) 297.48
MDL Number MFCD00149889
InChI Key JGPSMWXKRPZZRG-UHFFFAOYSA-N
Synonym zinc nitrate hexahydrate, zinc ii nitrte, zincnitratehexahydrate, acmc-1bzdd, n2o6zn.6h2o, ksc176g7t, zinc nitrate hexahydrate, puratronic, zinc nitrate, zinc 2+ hexahydrate dinitrate, zinc nitrate hexahydrate, p.a
PubChem CID 15865313
IUPAC Name zinc;dinitrate;hexahydrate
SMILES O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 36°C
Color White
pH 5.1
Physical Form Solid
Quantity 250 g
Formula Weight 297.46g/mol
Packaging Plastic powder jar
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'