missing translation for 'onlineSavingsMsg'
Learn More

Zinc neodecanoate, Zn 17.9-18.2%

Product Code. 11389677
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11389677 1 kg 1kg
11309687 250 g 250g
11399677 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11389677 Supplier Thermo Scientific Alfa Aesar Supplier No. 041312.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 27253-29-8 | C20H40O4Zn | 409.916 g/mol

Zinc neodecanoate is used as a catalyst, lubricant additive and drier. Further it is used in the production of rubber and plastic products. In addition it serves as a polyurethane catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27253-29-8
Molecular Formula C20H40O4Zn
Molecular Weight (g/mol) 409.916
MDL Number MFCD00014384
InChI Key VADFQXONMAETJP-UHFFFAOYSA-N
Synonym zinc neodecanoate, zinc neodecanoate, zn, 7,7-dimethyloctanoyl oxy zincio 7,7-dimethyloctanoate
PubChem CID 117064711
IUPAC Name 7,7-dimethyloctanoic acid;zinc
SMILES CC(C)(C)CCCCCC(=O)O.CC(C)(C)CCCCCC(=O)O.[Zn]

Specifications

Color Brown
Flash Point 102°C (216°F)
Quantity 1 kg
Solubility Information Immiscible with water.
Formula Weight 407.89
Physical Form Liquid
Chemical Name or Material Zinc neodecanoate
Product Identifier
  • Zinc neodecanoate
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Neodecanoic acid, zinc salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.