missing translation for 'onlineSavingsMsg'
Learn More

Zinc naphthenate, ^=67% in mineral spirits (10% Zn)

Product Code. p-7050824
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
This item is not returnable. View return policy

Product Code. 11318368

Brand: Thermo Scientific Alfa Aesar 039554.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12001-85-3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12001-85-3
MDL Number MFCD00147699

Specifications

Color Colorless
Density 1.01 g/mL
Flash Point >37.8° (100°F)
Quantity 500 g
UN Number UN1993
Physical Form Liquid
Chemical Name or Material Zinc naphthenate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.