missing translation for 'onlineSavingsMsg'
Learn More

Zinc molybdenum oxide, 98%

Product Code. 11413870
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413870 5 g 5g
11423870 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11413870 Supplier Thermo Scientific Alfa Aesar Supplier No. 089937.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13767-32-3 | ZnMoO4

Used as a corrosion inhibitor in paints, adhesives, primer on metal surfaces, such as steel and aluminum. It may be applied by brush or spray, and is suitable as an after-blast or after-pickling, primer for plating, as an undercoat for alkyd enamels. Also used for dielectric and insulation applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13767-32-3
Molecular Formula ZnMoO4
MDL Number MFCD00049622
Synonym Zinc molybdate

Specifications

Melting Point >700°C
Physical Form Powder
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 225.32
Percent Purity 98%
Chemical Name or Material Zinc molybdenum oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.