missing translation for 'onlineSavingsMsg'
Learn More

Zinc Isotopic Standard: Zn @ 10 μg/mL in 2% HNO3 (Enriched in Zn-68)

Product Code. 30323883
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
Product Code. Quantity unitSize
30323883 50 mL 50mL
1 options
This item is not returnable. View return policy

Product Code. 30323883

Brand: VHG Labs VHGLIS68ZN50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Zinc-68
CAS 14378-35-9
Analyte or Component Names Zinc-68
Shipping Condition Room Temperature
Molecular Formula 68 Zn
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Synonym 68 Zn
InChI Formula InChI=1 S/Zn/i1+3
Solution Type Single Solution
SMILES [68 Zn]
Element Zn
IUPAC Name zinc-68
Solvent or Matrix Dilute HNO3
Industry Type Petrochemical Testing
Molecular Weight (g/mol) 67.92
Quantity 50 mL
Formula Weight 67.9248
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.