Learn More
Zinc iron oxide
Description
Zinc iron oxide is used as pigments, especially in applications related to heat stability. It is also used in high corrosion-resistant coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Fine Powder |
| Quantity | 100 g |
| Solubility Information | Insoluble in water. |
| Formula Weight | 241.06 |
| Odor | Odorless |
| Chemical Name or Material | Zinc iron oxide |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.