missing translation for 'onlineSavingsMsg'
Learn More

Zinc iodide, ultra dry, 99.995% (metals basis)

Product Code. 11388587
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388587 25 g 25g
11398587 5 g 5g
11308597 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11388587 Supplier Thermo Scientific Alfa Aesar Supplier No. 035727.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10139-47-6 | ZnI2

Zinc iodide is used as an x-ray opaque penetrant in industrial radiography to enhance the contrast. It is used as electrode compartments and as a stain in electron microscopy in association with osmium tetroxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10139-47-6
Molecular Formula ZnI2
MDL Number MFCD00011299

Specifications

Boiling Point 625°C (decomposition)
Melting Point 446°C
Physical Form Granules
Quantity 25 g
Assay Percent Range 99.9%, metal basis
Solubility Information Soluble in water.
Formula Weight 319.18
Sensitivity Hygroscopic
Density 4.74
Chemical Name or Material Zinc Iodide, Ampouled under argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.