missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Iodide, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Zinc Iodide |
| CAS | 10139-47-6 |
| Molecular Formula | I2Zn |
| Quantity | 1 g |
| Synonym | NSC 39113, Zinc Diiodide |
| InChI Formula | InChI=1S/2HI.Zn/h2*1H;/q;;+2/p-2 |
| SMILES | I[Zn]I |
| IUPAC Name | diiodozinc |
| Molecular Weight (g/mol) | 319.19 |
| Formula Weight | 317.7381 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction