missing translation for 'onlineSavingsMsg'
Learn More

Zinc iodide, 99.999%, (trace metal basis), extra pure

Product Code. 10378010 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10378010 25 g Glass bottle 25g
10346591 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10378010 Supplier Thermo Scientific Acros Supplier No. 212760250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10139-47-6 | I2Zn | 319.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10139-47-6
Molecular Formula I2Zn
Molecular Weight (g/mol) 319.19
MDL Number MFCD00011299

Specifications

Melting Point 446°C
Quantity 25 g
Assay Percent Range 99.999%
Linear Formula ZnI2
Fieser 01,1294; 05,765; 10,462; 11,604; 12,576; 13,88
Merck Index 15, 10339
Solubility Information Solubility in water: 4500g/L (20°C). Other solubilities: soluble in alkalies,soluble in alcohol and ether
Formula Weight 319.19
Percent Purity 100.00%
Grade Trace Metal Basis
Packaging Glass bottle
Specific Gravity 4.74
Density 4.7400g/mL
Chemical Name or Material Zinc iodide, 99.999%
Show More Show Less
compliance-icons
Product Identifier
  • Zinc iodide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Zinc iodide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.