missing translation for 'onlineSavingsMsg'
Learn More

Zinc ingot, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number 15420967
Click to view available options
Quantity:
1 Pc.
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS 7440-66-6
Molecular Formula Zn
MDL Number MFCD00011291
Quantity 1 Pc.
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
SMILES [Zn]
IUPAC Name zinc
Molecular Weight (g/mol) 65.38
PubChem CID 23994
ChEBI CHEBI:30185
Percent Purity 99.9999%
Odor Odorless
Weight ≈200g/ingot
Form Ingot
Assay (metals basis)
Chemical Name or Material Zinc ingot
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'