missing translation for 'onlineSavingsMsg'
Learn More

Zinc(II) Trifluoromethanesulphonate, TRC

Product Code. 30449777
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30449777 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30449777 Supplier Toronto Research Chemicals Supplier No. TRCZ439683500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Zinc(II) Trifluoromethanesulphonate
CAS 54010-75-2
Molecular Formula C F3 O3 S . Zn
Quantity 500 mg
InChI Formula InChI=1S/CHF3O3S.Zn/c2-1(3,4)8(5,6)7;/h(H,5,6,7);/q;+2/p-1
SMILES [Zn+2].[O-]S(=O)(=O)C(F)(F)F
IUPAC Name zinc;trifluoromethanesulfonate
Molecular Weight (g/mol) 214.478
Formula Weight 212.881
Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.