missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Hydroxide, MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| CAS | 20427-58-1 |
| Quantity | 100 g |
| Molecular Formula | H2O2Zn |
| Synonym | zinc hydroxide, zinc hydroxide zn oh 2, zinc 2+ ion dioxidanide, zinc, ion zn2+ bis oh- |
| InChI Key | UGZADUVQMDAIAO-UHFFFAOYSA-L |
| SMILES | [OH-].[OH-].[Zn+2] |
| IUPAC Name | zinc;dihydroxide |
| Molecular Weight (g/mol) | 99.394 |
| PubChem CID | 9812759 |
| Formula Weight | 99.38 |
| Show More |
Safety and Handling
Signal Word
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P264-Wash thoroughly after handling.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Zinc Hydroxide
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction