missing translation for 'onlineSavingsMsg'
Learn More

Zinc, Honeywell Fluka™

Product Code. 16655773 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
This item is not returnable. View return policy

Product Code. 16655773

Brand: Honeywell Fluka 243469H100G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Granular, 20-30 mesh, ACS reagent, ≥99.8%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-66-6
PubChem CID 23994
ChEBI CHEBI:30185

Specifications

Physical Form Granules
Quantity 100 g
Percent Purity ≥99.80%
Grade ACS
Packaging Plastic Bottle
Appearance Complying
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.