missing translation for 'onlineSavingsMsg'
Learn More

Zinc granules, ACS, -20 mesh, 99.8% min

Product Code. p-4408698
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
This item is not returnable. View return policy

Product Code. 11328048

Brand: Thermo Scientific Alfa Aesar 036602.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Zinc Granules; ACS; 99.8%; Alfa Aesar; -20 Mesh | CAS: 7440-66-6 | Zn | 65.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]

Specifications

Quantity 100 g
Mesh Size -20 Mesh
Assay Percent Range 99.8%
Form Granular
Grade ACS Reagent
Chemical Name or Material Zinc, For analysis ACS

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.