missing translation for 'onlineSavingsMsg'
Learn More

Zinc gluconate hydrate, 97%, Thermo Scientific Chemicals

Product Code. 11416206
Click to view available options
Quantity:
100 g
500 g

Zinc gluconate hydrate is used as a food additive, a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc gluconate hydrate is used as a food additive, a pharmaceutical intermediate.

Solubility
Soluble in water 100g/ l.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4468-02-4
MDL Number MFCD00868110

Specifications

pH 6.5
Odor Odorless
Molecular Formula C4H7NO
Quantity 500 g
Merck Index 14,4456
Synonym propionaldehyde cyanohydrin, 2-hydroxybutyronitrile, butyronitrile, 2-hydroxy, butanenitrile, 2-hydroxy, alpha-hydroxybutyronitrile, propanal cyanhydrin, 1-cyanopropan-1-ol, 2-oxidanylbutanenitrile, 2-hydroxy-butyronitrile, butanenitrile, 2-hydroxy-9ci
Solubility Information Soluble in water 100g/ l.
InChI Key NHSSTOSZJANVEV-UHFFFAOYNA-N
SMILES CCC(C#N)O
IUPAC Name 2-hydroxybutanenitrile
Molecular Weight (g/mol) 85.11
PubChem CID 20553
Formula Weight 455.69 (Anhydrous)
Percent Purity 97%
Chemical Name or Material Zinc gluconate hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'