missing translation for 'onlineSavingsMsg'
Learn More

Zinc fluoride tetrahydrate, 98%, Thermo Scientific Chemicals

Product Code. 11357727
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357727 100 g 100g
11347727 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357727 Supplier Thermo Scientific Alfa Aesar Supplier No. 014127.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Zinc fluoride tetrahydrate is used as a mild fluorinating agent in the replacement of chlorine in halogenated hydrocarbons, in fluorophosphate and fluoride glass, and in the manufacture of fluoride glass optical fibers and optical transmitting glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13986-18-0
Molecular Formula F2H8O4Zn
Molecular Weight (g/mol) 175.437
MDL Number MFCD00149892
InChI Key OEUUYPRXVRBIFO-UHFFFAOYSA-L
Synonym zinc fluoride tetrahydrate, zinc fluoride, tetrahydrate, unii-sq7815wxfb, zinc difluoride tetrahydrate, sq7815wxfb, zinc fluoride tetrahydrate mi, acmc-1c1qo, ksc497c6d, zinc, ion zn2+ tetrahydrate difluoride, zinc fluoride, tetrahydrate,
PubChem CID 44717730
IUPAC Name zinc;difluoride;tetrahydrate
SMILES O.O.O.O.[F-].[F-].[Zn+2]

Specifications

Physical Form Crystalline
Quantity 100 g
UN Number UN3288
Merck Index 14,10136
Solubility Information Slightly soluble in water
Formula Weight 175.43 (103.37 Anhydrous)
Percent Purity 98%
Odor Odorless
Chemical Name or Material Zinc fluoride tetrahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.