missing translation for 'onlineSavingsMsg'
Learn More

Zinc fluoride, Puratronic™, 99.995% (metals basis)

Product Code. 15466005
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15466005 100 g 100g
11302088 5 g 5g
11312088 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15466005 Supplier Thermo Scientific Alfa Aesar Supplier No. 012986.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7783-49-5 | F2Zn | 103.377 g/mol

It is widely used in fluorination of organic compounds, as additive of electrolytic aluminum and also in electroplating baths. It is used in preserving food, manufacture of flux, oxygen-sensitive applications such as metal production, glasses, enamels, fabrics and plastics. It is also used in oil refining, manufacture of pharmaceuticals, manufacture phosphors for fluorescent electric lights, and floor coverings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-49-5
Molecular Formula F2Zn
Molecular Weight (g/mol) 103.377
MDL Number MFCD00011298
InChI Key BHHYHSUAOQUXJK-UHFFFAOYSA-L
Synonym zinc ii fluoride, zinc 2+ ion difluoride, acmc-1bhy1, ksc379c0l, zinc fluoride 50g, zinc, ion zn2+ difluoride
PubChem CID 10129912
IUPAC Name zinc;difluoride
SMILES [F-].[F-].[Zn+2]

Specifications

Boiling Point 1500°C
Melting Point 872°C
Color White
Physical Form Powder
Quantity 100 g
Linear Formula ZnF2
Merck Index 14,10136
Solubility Information Sparingly soluble in hydrochloric acid,nitric acid,and ammonium hydroxide.
Formula Weight 103.37
Percent Purity 99.995%
Grade Puratronic
Density 4.95 g/mL
Assay (metals basis)
Chemical Name or Material Zinc fluoride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.