missing translation for 'onlineSavingsMsg'
Learn More

Zinc Fluoride, 99%, Honeywell Fluka™

Product Code. 15661770 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15661770 500 g Poly Bottle 500g
15651770 25 g Poly Bottle 25g
15641770 100 g Poly Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15661770 Supplier Honeywell-Fluka Supplier No. 205567500G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Powder, 99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-49-5
Molecular Formula F2Zn
Molecular Weight (g/mol) 103.377
MDL Number MFCD00011298
InChI Key BHHYHSUAOQUXJK-UHFFFAOYSA-L
Synonym zinc ii fluoride, zinc 2+ ion difluoride, acmc-1bhy1, ksc379c0l, zinc fluoride 50g, zinc, ion zn2+ difluoride
PubChem CID 10129912
IUPAC Name zinc;difluoride
SMILES [F-].[F-].[Zn+2]

Specifications

Melting Point 872°C
Color White
Physical Form Powder
Quantity 500 g
Linear Formula ZnF2
Formula Weight 103.39g/mol
Percent Purity 99%
Packaging Poly Bottle
Vapor Pressure 1mmHg (970°C)
Density 4.95g/mL at 25°C
Chemical Name or Material Zinc Fluoride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.