missing translation for 'onlineSavingsMsg'
Learn More

Zinc fluoride, 98%

Product Code. 10510362 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
250 g
Packaging:
Plastic bottle
Unit Size:
250g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10510362 250 g Plastic bottle 250g
10009061 25 g Plastic bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10510362 Supplier Thermo Scientific Acros Supplier No. 363912500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7783-49-5 | F2Zn | 103.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-49-5
Molecular Formula F2Zn
Molecular Weight (g/mol) 103.38
MDL Number MFCD00011298
InChI Key BHHYHSUAOQUXJK-UHFFFAOYSA-L
Synonym zinc ii fluoride, zinc 2+ ion difluoride, acmc-1bhy1, ksc379c0l, zinc fluoride 50g, zinc, ion zn2+ difluoride
PubChem CID 10129912
IUPAC Name zinc;difluoride
SMILES [F-].[F-].[Zn+2]

Specifications

Boiling Point 1500.0°C
Melting Point 872.0°C to 910.0°C
Color Brown to White
Physical Form Crystals or Powder
Quantity 250 g
Assay Percent Range 98%
Linear Formula ZnF2
Fieser 06,514
Merck Index 15, 10335
Solubility Information Solubility in water: slightly soluble. Other solubilities: freely soluble in hcl, hno3 and nh4oh, insoluble in alcohol
Formula Weight 103.38
Percent Purity 98%
Packaging Plastic bottle
Chemical Name or Material Zinc fluoride, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.