missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc finger protein 668 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-86427-0.1ML
This item is not returnable.
View return policy
Description
Zinc finger protein 668 Polyclonal specifically detects Zinc finger protein 668 in Human samples. It is validated for Western Blot.
Specifications
| Zinc finger protein 668 | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 79759 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| FLJ13479, FLJ14492, zinc finger protein 668 | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of human Zinc finger protein 668 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering