missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc finger protein 639 Antibody (PCRP-ZNF639-2B2), DyLight 550, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Zinc finger protein 639 Monoclonal antibody specifically detects Zinc finger protein 639 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Specifications
Specifications
| Antigen | Zinc finger protein 639 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Clone | PCRP-ZNF639-2B2 |
| Conjugate | DyLight 550 |
| Dilution | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | ANC_2H01, ZASC1ANC-2H01, zinc finger amplified in esophageal squamous cell carcinomas 1, zinc finger protein 639, Zinc finger protein ANC_2H01, Zinc finger protein ZASC1,6230400O18Rik |
| Host Species | Mouse |
| Immunogen | Recombinant human fragment (aa 406-485) of Zinc finger protein 639 (Uniprot: Q9UID6) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?