missing translation for 'onlineSavingsMsg'
Learn More

Zinc finger protein 639 Antibody (PCRP-ZNF639-2B2), Biotin, Novus Biologicals™

Product Code. 18393725 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18393725 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18393725 Supplier Novus Biologicals Supplier No. NBP314131B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Zinc finger protein 639 Monoclonal antibody specifically detects Zinc finger protein 639 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Zinc finger protein 639
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone PCRP-ZNF639-2B2
Conjugate Biotin
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias ANC_2H01, ZASC1ANC-2H01, zinc finger amplified in esophageal squamous cell carcinomas 1, zinc finger protein 639, Zinc finger protein ANC_2H01, Zinc finger protein ZASC1,6230400O18Rik
Host Species Mouse
Immunogen Recombinant human fragment (aa 406-485) of Zinc finger protein 639 (Uniprot: Q9UID6)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51193
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.