missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc finger protein 395 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-10967-100UL
This item is not returnable.
View return policy
Description
Zinc finger protein 395 Polyclonal specifically detects Zinc finger protein 395 in Human samples. It is validated for Western Blot.
Specifications
| Zinc finger protein 395 | |
| Polyclonal | |
| Western Blot 1.0 ug/ml | |
| dJ874C20.2, FLJ23407, zinc finger protein 310 pseudogene, zinc finger protein 323, ZNF20-Lp, ZNF310P | |
| The immunogen is a synthetic peptide directed towards the C terminal region of human Zinc finger protein 395 (NP_061130). Peptide sequence GLPLSALPPPLHKAQSSGPEHPGPESSLPSGALSKSAPGSFWHIQADHAY | |
| 100 μg | |
| DNA Repair, DNA replication Transcription Translation and Splicing | |
| 55893 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| PBS buffer, 2% sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction