missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc finger protein 287 Antibody (3A2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00057336-M08
This item is not returnable.
View return policy
Description
Zinc finger protein 287 Monoclonal antibody specifically detects Zinc finger protein 287 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA
Specifications
| Zinc finger protein 287 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| MGC15716, Zfp414, zinc finger protein 414 | |
| ZNF287 (NP_065704.1, 231 a.a. ∽ 330 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. PEWETKAQACTPVEDMSKLTKEETHTIKLEDSYDYDDRLERRGKGGFWKIHTDERGFSLKSVLSQEYDPTEECLSKYDIYRNNFEKHSNLIVQFDTQLDN | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunofluorescence, Sandwich ELISA | |
| 3A2 | |
| Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA | |
| NP_065704 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 57336 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction