missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Zinc finger and BTB domain-containing protein 9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29120
This item is not returnable.
View return policy
Description
Zinc finger and BTB domain-containing protein 9 Polyclonal antibody specifically detects Zinc finger and BTB domain-containing protein 9 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| Zinc finger and BTB domain-containing protein 9 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| MGC23166, zinc finger and BTB domain containing 9, zinc finger and BTB domain-containing protein 9, ZNF919 | |
| between 250 and 300 | |
| 100 μg | |
| Cancer, Cell Cycle and Replication | |
| 221504 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction