missing translation for 'onlineSavingsMsg'
Learn More

Zinc cyclohexanebutyrate dihydrate, AAS, Thermo Scientific™

Product Code. 11354697
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354697 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11354697 Supplier Thermo Scientific Alfa Aesar Supplier No. 039734.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38582-18-2
Molecular Formula C20H34O4Zn
Molecular Weight (g/mol) 403.87
MDL Number MFCD00035537
InChI Key KVSXXOUZYJZCPY-UHFFFAOYSA-L
Synonym zinc cyclohexylbutyrate, cyclohexanebutanoic acid, zinc salt, zinc cyclohexanebutyrate, cyclohexanebutanoic acid, zinc salt 2:1, zinc bis 4-cyclohexylbutanoate, zinc 4-cyclohexylbutanoate, acmc-1bn8b, bis cyclohexanebutyric acid zinc salt
PubChem CID 170080
IUPAC Name zinc;4-cyclohexylbutanoate
SMILES [Zn++].[O-]C(=O)CCCC1CCCCC1.[O-]C(=O)CCCC1CCCCC1

Specifications

Melting Point 77°C to 80°C
Quantity 5 g
Formula Weight 439.91 (403.87 Anhydrous)
Physical Form Powder
Chemical Name or Material Zinc cyclohexanebutyrate dihydrate, AAS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.