missing translation for 'onlineSavingsMsg'
Learn More

Zinc Cyanide, TRC

Product Code. 30456106
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456106 50 g 50g
30450361 25 g 25g
30451278 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456106 Supplier Toronto Research Chemicals Supplier No. TRCZ43950050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 557-21-1
Molecular Formula C2N2Zn
Molecular Weight (g/mol) 117.41
Synonym Dicyanozinc, Zinc Dicyanide
IUPAC Name zinc;dicyanide
SMILES [Zn+2].[C-]#N.[C-]#N

Specifications

Quantity 50 g
InChI Formula InChI=1S/2CN.Zn/c2*1-2;/q2*-1;+2
Formula Weight 115.9353
Chemical Name or Material Zinc Cyanide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.