missing translation for 'onlineSavingsMsg'
Learn More

Zinc cyanide, 98%

Product Code. 11690222
Change view
Click to view available options
Quantity:
25 g
100 g
1000 g
Unit Size:
1000g
100g
1kg
25g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11690222 100 g 100g
11466840 25 g 25g
11456840 1000 g 1000g
11332797 1000 g 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11690222 Supplier Thermo Scientific Alfa Aesar Supplier No. L15836.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 557-21-1 | Zn(CN)2

Zinc cyanide is used for electroplating of zinc. In Gatterman reaction , it is used for the introduction of formyl group in to aromatic compounds as well as to prepare 2-hydroxy-1-nafthaldehyde and mesitaldehyde. It is also used in chemical analysis in analytical chemistry. Further, it acts as a catalyst for the cyanosilylation of aldehydes and ketones. In addition to this, it reacts with 1,3,5-triethyl-benzene to get 2,4,6-triethyl-benzaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc cyanide is used for electroplating of zinc. In Gatterman reaction , it is used for the introduction of formyl group in to aromatic compounds as well as to prepare 2-hydroxy-1-nafthaldehyde and mesitaldehyde. It is also used in chemical analysis in analytical chemistry. Further, it acts as a catalyst for the cyanosilylation of aldehydes and ketones. In addition to this, it reacts with 1,3,5-triethyl-benzene to get 2,4,6-triethyl-benzaldehyde.

Solubility
Soluble in alkalies, potassium cyanide and ammonia. Insoluble in water and most solvents.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 557-21-1
Molecular Formula Zn(CN)2
MDL Number MFCD00011292

Specifications

Melting Point 800°C (decomposition)
Quantity 100 g
Linear Formula Zn(CN)2
UN Number UN1713
Beilstein 4124366
Merck Index 14,10135
Solubility Information Soluble in alkalies,potassium cyanide and ammonia. Insoluble in water and most solvents.
Formula Weight 117.42
Percent Purity 98%
Sensitivity Hygroscopic
Density 1.852
Chemical Name or Material Zinc cyanide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.