missing translation for 'onlineSavingsMsg'
Learn More

Zinc cyanide, 85%

Product Code. 11386469
Change view
Click to view available options
Quantity:
100 g
4 x 500 g
Unit Size:
100g
2000g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11386469 4 x 500 g 2000g
11366469 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11386469 Supplier Thermo Scientific Alfa Aesar Supplier No. 014158.57

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Electroplating, reagent | CAS: 557-21-1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 557-21-1
MDL Number MFCD00011292

Specifications

Melting Point 800°C (decomposition)
Color White
Physical Form Crystalline
Quantity 4 x 500 g
UN Number UN1713
Beilstein 4124366
Merck Index 14,10135
Solubility Information Insoluble in water, alcohol; Soluble in solutions of alkali cyanides or hydroxides
Formula Weight 117.42
Percent Purity 85%
Sensitivity Hygroscopic
Density 1.852 g/mL
Chemical Name or Material Zinc cyanide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.