missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc-Copper Couple, TRC
Click to view available options
Quantity:
50 g
Unit Size:
50g
Specifications
Specifications
| Chemical Name or Material | Zinc-Copper Couple |
| CAS | 53801-63-1 |
| Molecular Formula | CuZn |
| Quantity | 50 g |
| Synonym | Copper','compd. with zinc |
| InChI Formula | InChI=1S/Cu.Zn |
| SMILES | [Cu].[Zn] |
| IUPAC Name | copper;zinc |
| Molecular Weight (g/mol) | 128.93 |
| Formula Weight | 126.8587 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion