missing translation for 'onlineSavingsMsg'
Learn More

Zinc-copper couple, Copper content typically ca 1-4%

Product Code. p-7045203
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11312277

Brand: Thermo Scientific Alfa Aesar L09811.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 53801-63-1 | CuZn | 128.93 g/mol

Zinc-copper couple is used in the Simmons-Smith cyclopropanation reaction, Reformatsky reaction. It is used as a chemical for the selective, stereospecific reduction of alkynes to cis-alkenes. It is used in for deiodination of iodotributylstannylalkanes. In the presence of a free-radical initiator, it promotes the formylation of perfluoroalkyl iodides with DMF to give fluorinated aldehydes in high yield. It is used in dermatology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc-copper couple is used in the Simmons-Smith cyclopropanation reaction, Reformatsky reaction. It is used as a chemical for the selective, stereospecific reduction of alkynes to cis-alkenes. It is used in for deiodination of iodotributylstannylalkanes. In the presence of a free-radical initiator, it promotes the formylation of perfluoroalkyl iodides with DMF to give fluorinated aldehydes in high yield. It is used in dermatology.

Solubility
Insoluble in water.

Notes
Store at 4°C. Moisture Sensitive. Do not store together with acids. Protect from humidity and water. Incompatible materials are amines, cadmium, chlorinated solvents, Sulfur compounds.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53801-63-1
Molecular Formula CuZn
Molecular Weight (g/mol) 128.93
MDL Number MFCD00084851
InChI Key TVZPLCNGKSPOJA-UHFFFAOYSA-N
Synonym zinc-copper couple, cu.zn, zinc copper, copper, compound with zinc 1:1, zinc-copper, brass alloy, zn copper, copper-zinc alloy, zinc copper couple, cu-zn
PubChem CID 10290809
IUPAC Name copper zinc
SMILES [Cu].[Zn]

Specifications

Melting Point 419°C to 420°C
Quantity 25 g
Sensitivity Moisture sensitive
UN Number UN3089
Solubility Information Insoluble in water.
Assay Copper content typically ≈1-3%
Chemical Name or Material Zinc-copper couple

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.