Learn More
Zinc Copper composite, -100 mesh, 99% (metals basis), Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 044013.09
14.06 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg Transport : UN number : 3089 Chem class : 4.1 Pack group : III
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Zinc Copper composite | |
Typically Zn 90 to 95, Cu 5 to 10 wt% | |
MFCD03102068 | |
10 g | |
−100 Mesh |
Alloy Precursor | |
(metals basis) | |
UN3089 | |
99% |
RUO – Research Use Only