missing translation for 'onlineSavingsMsg'
Learn More

Zinc chromate

Product Code. 11473697
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
This item is not returnable. View return policy

Product Code. 11473697

Brand: Thermo Scientific Alfa Aesar A18178.0B

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13530-65-9 | CrO4Zn | 181.37 g/mol

Zinc chromate is used in pigments, rubber, linoleum manufacturing, metal surface treatment, electroplating intermediates, research and development. It finds its application as an useful metal for preventing corrosion when applied to other metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc chromate is used in pigments, rubber, linoleum manufacturing, metal surface treatment, electroplating intermediates, research and development. It finds its application as an useful metal for preventing corrosion when applied to other metals.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13530-65-9
Molecular Formula CrO4Zn
Molecular Weight (g/mol) 181.37
MDL Number MFCD00036281
InChI Key NDKWCCLKSWNDBG-UHFFFAOYSA-N
Synonym zinc chromate, zinc chrome, zinc chromate c, zinc chromate o, zinc chromate t, zinc chromate z, zinc chromate am, zincro zto, basic zinc chromate, zinc chrome yellow
PubChem CID 26089
IUPAC Name zinc;dioxido(dioxo)chromium
SMILES [Zn++].[O-][Cr]([O-])(=O)=O

Specifications

Density 3.4
Quantity 1000 g
UN Number UN3077
Solubility Information Insoluble in water.
Formula Weight 181.37
Chemical Name or Material Zinc chromate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.