missing translation for 'onlineSavingsMsg'
Learn More

Zinc chloride, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number 11368587
Click to view available options
Quantity:
5 g
25 g
100 g
7646-85-7
Cl2Zn
136.28
MFCD00011295
JIAARYAFYJHUJI-UHFFFAOYSA-L
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
5727
CHEBI:49976
dichlorozinc
Cl[Zn]Cl
7646-85-7
Cl2Zn
136.28
MFCD00011295
JIAARYAFYJHUJI-UHFFFAOYSA-L
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
5727
CHEBI:49976
dichlorozinc
Cl[Zn]Cl

Zinc(II) chloride is used as a catalyst, as a dehydrating and condensing agent, soldering flux and metal etchant. It is also employed in preserving anatomical specimens, wood preservatives, deodorant, disinfecting and embalming materials. It is employed as a mordant in printing and dyeing materials, and in the vulcanizing processes of fiber and rubber. It is useful as an electrolyte in dry cell batteries, in metal industry, in galvanizing iron and as an electrolyte for electroplating. It is useful in an organic synthesis, for example in the Friedel-crafts acylation, Fisher indole synthesis, Lucas reagent (zinc chloride with HCl), activation of allylic/benzylic halides towards reaction with olefins or with sodium cyanoborohydride catalyzed reduction of halides, organozinc reagents for use in Negishi coupling, and aldol condensation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 732°C
Melting Point 290°C
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.999% (metals basis)
Solubility Information Freely soluble in water and dilute hydrochloric acid. Soluble in alcohol,glycerol,ether and acetone.
Formula Weight 136.29
Sensitivity Hygroscopic
Density 2.91 g/mL
Chemical Name or Material Zinc chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'