missing translation for 'onlineSavingsMsg'
Learn More

Zinc Chloride Solution, Honeywell™

Product Code. 15652920 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mL
Packaging:
Glass Bottle
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 15652920

Brand: Honeywell-Fluka 276839100ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 7646-85-7
Quantity 100 mL
Molecular Formula Cl2Zn
Linear Formula ZnCl2
MDL Number MFCD00011295
UN Number UN2924
Flash Point −40°C (−40°F)
Synonym zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
SMILES Cl[Zn]Cl
IUPAC Name dichlorozinc
Molecular Weight (g/mol) 136.28
PubChem CID 5727
ChEBI CHEBI:49976
Formula Weight 136.3g/mol
Packaging Glass Bottle
Density 0.835g/mL at 25°C
Chemical Name or Material Zinc Chloride Solution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.