missing translation for 'onlineSavingsMsg'
Learn More

Zinc Chloride Solution, 0.5M in Tetrahydrofuran, Honeywell™

Product Code. 15655130 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
Glass Bottle
Unit Size:
100mL
800mL
This item is not returnable. View return policy

Product Code. 15655130

Brand: Honeywell-Fluka 366374100ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

0.5 M in Tetrahydrofuran

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-85-7
Molecular Formula Cl2Zn
Molecular Weight (g/mol) 136.28
MDL Number MFCD00011295
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
Synonym zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
PubChem CID 5727
ChEBI CHEBI:49976
IUPAC Name dichlorozinc
SMILES Cl[Zn]Cl

Specifications

Color Colorless
Quantity 100 mL
Linear Formula ZnCl2
UN Number UN1993BF
Flash Point −20°C (−4°F)
Formula Weight 136.3g/mol
Packaging Glass Bottle
Appearance Liquid
Density 0.948g/mL at 25°C
Chemical Name or Material Zinc chloride solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.