missing translation for 'onlineSavingsMsg'
Learn More

Zinc chloride hydrate, Puratronic™, 99.999% (metals basis)

Product Code. 11338677
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338677 250 g 250g
11318677 10 g 10g
11328677 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11338677 Supplier Thermo Scientific Alfa Aesar Supplier No. 035782.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 29604-34-0 | Cl2Zn | 136.28 g/mol

It is used for a source of the zinc ion. It can also be used alone or with phenols for preserving railway ties, fireproofing timber, and with ammonium chloride as a flux for soldering and etching of metals. It is used as a moderate-strength lewis acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29604-34-0
Molecular Formula Cl2Zn
Molecular Weight (g/mol) 136.28
MDL Number MFCD00011295
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
Synonym zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
PubChem CID 5727
ChEBI CHEBI:49976

Specifications

Color White
Physical Form Crystalline Aggregates
Quantity 250 g
Linear Formula ZnCl2·xH2O
UN Number UN3260
Merck Index 14,10132
Solubility Information Soluble in water,ethanol (slightly),acetone (slightly),ether,and glycerol.
SMILES [Cl-].[Cl-].[Zn++]
IUPAC Name zinc(2+) dichloride
Formula Weight 136.29 (Anhydrous)
Percent Purity 99.999%
Grade Puratronic
Odor Odorless
Assay (metals basis)
Chemical Name or Material Zinc chloride hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.