missing translation for 'onlineSavingsMsg'
Learn More

Zinc chloride hydrate, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog Number 11319627
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
29604-34-0
Cl2Zn
136.28
MFCD00011295
JIAARYAFYJHUJI-UHFFFAOYSA-L
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
5727
CHEBI:49976
dichlorozinc
Cl[Zn]Cl
This item is not returnable. View return policy
29604-34-0
Cl2Zn
136.28
MFCD00011295
JIAARYAFYJHUJI-UHFFFAOYSA-L
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
5727
CHEBI:49976
dichlorozinc
Cl[Zn]Cl

Zinc(II) chloride hydrate is used as a soldering flux and metal etchant. It is also employed in preserving anatomical specimens, wood preservatives, deodorant, disinfecting and embalming materials. It is employed as a mordant in printing and dyeing materials, and in the vulcanizing processes of fiber and rubber. It forms a complex with Ruhemann’s purple, which obtained by reacting ninhydrin with amino acids and amines is used in fingerprint detection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Crystalline Aggregates
Quantity 10 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Soluble in water.
Formula Weight 136.29 (Anhydrous)
Chemical Name or Material Zinc chloride hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'