missing translation for 'onlineSavingsMsg'
Learn More

Zinc chloride, Honeywell

Product Code. 17278316 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
25 kg
Unit Size:
25kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17278316 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17278316 Supplier Honeywell Research Chemicals Supplier No. 7218625KG
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Melting Point 287°C to 304°C
CAS 7646-85-7
Color White
pH 5 (20°C)
Physical Form Solid
Quantity 25 kg
Molecular Formula Cl2Zn
Synonym Dichlorozinc
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
SMILES [Cl-].[Cl-].[Zn++]
IUPAC Name zinc(2+) dichloride
Molecular Weight (g/mol) 136.28
Percent Purity 100%
Packaging Paper Bag with Inliner
Chemical Name or Material Zinc chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.