missing translation for 'onlineSavingsMsg'
Learn More

Zinc Chloride, Free-flowing, Reagent Grade, anhydrous, ≥98%, Honeywell Fluka™

Product Code. 16635923
Change view
Click to view available options
Quantity:
25 kg
Unit Size:
25kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16635923 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16635923 Supplier Fluka Supplier No. 79352325KG
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 287° to 304°C
CAS 7646-85-7
Color White
CAS Max % 1
pH 5
Quantity 25 kg
Molecular Formula Cl2Zn
Linear Formula ZnCl2
Synonym Dichlorozinc
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
SMILES [Cl-].[Cl-].[Zn++]
IUPAC Name zinc(2+) dichloride
Molecular Weight (g/mol) 136.28
Density 2.93 g/cm3 (22°C)
Chemical Name or Material Zinc chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.