missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Chloride, Endotoxin Tested, Granular, Multi-Compendial, U.S.P., J.T. Baker™
370.50 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Specifications
Physical Form | Granules |
Quantity | 500 g |
Formula Weight | 136.3 |
Grade | Multi-Compendial/USP |
Packaging | Poly Bottle |
Specific Gravity | 1L = 2.91kg |
Chemical Name or Material | Zinc Chloride, Endotoxin Tested |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- Skin corrosion/irritation Category 1 B
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'