missing translation for 'onlineSavingsMsg'
Learn More

Zinc chloride, 2M solution in 2-MeTHF, AcroSeal™

Product Code. 10723342 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10723342 800 mL AcroSeal™ Glass bottle 800mL
10713342 100 mL AcroSeal™ Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10723342 Supplier Thermo Scientific Acros Supplier No. 428568000
  • ONLINE EXCLUSIVE PRICE
    £520.00
    £504.00 / 800mL
    Save £16.00
    3% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7646-85-7 | Cl2Zn | 136.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Zinc chloride
Name Note 2M solution in 2-Methyltetrahydrofuran
CAS 7646-85-7, 96-47-9
Color Colorless
Physical Form Liquid
Quantity 800 mL
Molecular Formula Cl2Zn
Linear Formula ZnCl2
MDL Number MFCD00011295
Flash Point −11°C
Merck Index 15, 10331
Synonym zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
SMILES Cl[Zn]Cl
IUPAC Name dichlorozinc
Molecular Weight (g/mol) 136.29
PubChem CID 5727
ChEBI CHEBI:49976
Formula Weight 136.29
Concentration or Composition (by Analyte or Components) 23 to 27%
Packaging AcroSeal™ Glass bottle
Specific Gravity 1.07
Density 1.0700g/mL
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Zinc chloride, 2M solution in 2-methyltetrahydrofuran
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.