missing translation for 'onlineSavingsMsg'
Learn More

Zinc chloride, 0.7M soln. in THF

Product Code. 15485747
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Product Code. Quantity unitSize
15485747 500 mL 500mL
15475747 100 mL 100mL
2 options
This item is not returnable. View return policy

Product Code. 15485747

Brand: Thermo Scientific Alfa Aesar H32962.AP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7646-85-7 | Cl2Zn | 136.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc chloride is a widely used as a moderate-strength Lewis acid. In biochemistry research this is used for a source of the zinc ion. It is also be used alone or with phenols for preserving railway ties, fireproofing timber, and with ammonium chloride as a flux for soldering and etching of metals. Zinc chloride is used in the synthesis of the ferrocenylarenes. It is used for the in situ generation of organozincate reagents by reacting with Grignard compounds and also to catalyze chloromethylation of resins for solid phase peptide synthesis.

Solubility
Soluble in water (4320 mg/ml at 25°C), ethanol (slightly), acetone (slightly), ether, and glycerol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture. Store under dry inert gas. Protect from humidity and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-85-7
Molecular Formula Cl2Zn
Molecular Weight (g/mol) 136.28
MDL Number MFCD00011295
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
Synonym zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
PubChem CID 5727
ChEBI CHEBI:49976
IUPAC Name dichlorozinc
SMILES Cl[Zn]Cl

Specifications

Quantity 500 mL
Linear Formula ZnCl2
UN Number UN1993
Solubility Information Soluble in water (4320mg/ml at 25°C),ethanol (slightly),acetone (slightly),ether,and glycerol.
Formula Weight 136.29
Concentration or Composition (by Analyte or Components) 0.7M soln. in THF
Density 0.948
Chemical Name or Material Zinc chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.