missing translation for 'onlineSavingsMsg'
Learn More

Zinc, Certified AR for Analysis, Powder (Metal), Fisher Chemical™

Product Code. 10774771 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10774771 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10774771 Supplier Fisher Chemical Supplier No. Z/0500/53
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]

Specifications

Color Gray
Melting Point 420°C
Boiling Point 907°C
Quantity 500 g
Packaging Plastic powder jar
Vapor Pressure 1.3mbar 478
Form Powder/Solid
Formula Weight 65.36g/mol
compliance-icons compliance-icons
Product Identifier
  • Zinc, powder
Signal Word
  • Danger
Hazard Category
  • Substance which\, in contact with water\, emits flammable gases Category 1
  • Pyrophoric solid Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H250-Catches fire spontaneously if exposed to air.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P222-Do not allow contact with air.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P370+P378-In case of fire: Use to extinguish.
  • P273-Avoid release to the environment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.