missing translation for 'onlineSavingsMsg'
Learn More

Zinc Carbonate, MP Biomedicals

Product Code. 11457591 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g

Product Code. 11457591

Brand: MP Biomedicals 0210473790

Please to purchase this item. Need a web account? Register with us today!

Chemical Identifiers

CAS 3486-35-9
Molecular Formula CO3Zn
Molecular Weight (g/mol) 125.388
InChI Key FMRLDPWIRHBCCC-UHFFFAOYSA-L
Synonym zinc carbonate, zincspar, carbonic acid, zinc salt 1:1, zinc monocarbonate, natural smithsonite, zinc carbonate 1:1, unii-k8290ptq4f, hydrozincite?, zinc carbonic acid, zinc 2+ ion carbonate
PubChem CID 19005
IUPAC Name zinc;carbonate
SMILES C(=O)([O-])[O-].[Zn+2]

Specifications

Quantity 500 g
Formula Weight 125.4
Chemical Name or Material Zinc Carbonate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.