missing translation for 'onlineSavingsMsg'
Learn More

Zinc carbonate basic, 97%, Zn >58.0%

Product Code. 11422008
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
This item is not returnable. View return policy

Product Code. 11422008

Brand: Thermo Scientific Alfa Aesar A14590.0B

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5263-02-5 | C2H6O12Zn5 | 548.96 g/mol

Zinc carbonate basic is used as pharmaceutical intermediate and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc carbonate basic is used as pharmaceutical intermediate and in chemical research.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5263-02-5
Molecular Formula C2H6O12Zn5
Molecular Weight (g/mol) 548.96
MDL Number MFCD00011519
InChI Key UOURRHZRLGCVDA-UHFFFAOYSA-D
Synonym Zinc subcarbonate; Zinc carbonate hydroxide
IUPAC Name pentazinc(2+) hexahydroxide dicarbonate
SMILES [OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Zn++].[Zn++].[Zn++].[Zn++].[Zn++].[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Quantity 1000 g
Solubility Information Insoluble in water.
Formula Weight 548.97
Percent Purity 97%
Assay Zn >58.0%
Chemical Name or Material Zinc carbonate basic
compliance-icons compliance-icons
Product Identifier
  • Zinc carbonate, basic
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Di[carbonato(2-)]hexahydroxypentazinc

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.