missing translation for 'onlineSavingsMsg'
Learn More

Zinc bromide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number 11348587
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
7699-45-8
Br2Zn
225.188
MFCD00011294
VNDYJBBGRKZCSX-UHFFFAOYSA-L
zincbromide, zinc ii bromide, acmc-209p7h, zinc 2+ ion dibromide, ksc378m6n, zinc bromide hydrate, puratronic™, zinc bromide, anhydrous 100g
10421208
zinc;dibromide
[Zn+2].[Br-].[Br-]
This item is not returnable. View return policy
7699-45-8
Br2Zn
225.188
MFCD00011294
VNDYJBBGRKZCSX-UHFFFAOYSA-L
zincbromide, zinc ii bromide, acmc-209p7h, zinc 2+ ion dibromide, ksc378m6n, zinc bromide hydrate, puratronic™, zinc bromide, anhydrous 100g
10421208
zinc;dibromide
[Zn+2].[Br-].[Br-]

Zinc bromide is an optimal catalyst for stereo specific and regioselective reaction of silacyclopropanes with carbonyl compounds. It is used as a transparent shield against radiation, as a fluid to displace drilling muds in transition phases, as a lewis acid in organic chemistry, colloidon emulsions for photography, a nutritional or functional ingredient in food and oral care products..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Boiling Point 650°C
Melting Point 394°C
Physical Form Beads
Quantity 5 g
UN Number UN3260
Merck Index 14,10129
Solubility Information Soluble in water,ethanol,ether,90% alcohol.
Formula Weight 225.19
Percent Purity 99.999%
Packaging Ampule
Refractive Index 1.5452
Sensitivity Hygroscopic
Density 4.201 g/mL
Assay (metals basis)
Chemical Name or Material Zinc bromide, Ultra dry
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'