missing translation for 'onlineSavingsMsg'
Learn More

Zinc bromide hydrate, 99.9% (metals basis)

Product Code. 11339748
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339748 500 g 500g
11329748 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11339748 Supplier Thermo Scientific Alfa Aesar Supplier No. 044230.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 299465-28-4 | Br2Zn | 225.19 g/mol

Zinc bromide is an optimal catalyst for stereospecific and regioselective reaction of silacyclopropanes with carbonyl compounds. Zinc bromide is employed as a lewis acid in organic chemistry. Has been used as a transparent shield against radiation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 299465-28-4
Molecular Formula Br2Zn
MDL Number MFCD00011294
InChI Key VNDYJBBGRKZCSX-UHFFFAOYSA-L
Synonym zincbromide, zinc ii bromide, acmc-209p7h, zinc 2+ ion dibromide, ksc378m6n, zinc bromide hydrate, puratronic™, zinc bromide, anhydrous 100g
PubChem CID 10421208

Specifications

Physical Form Crystalline
Quantity 500 g
UN Number UN3260
Merck Index 14,10129
Solubility Information Soluble in water (4470mg/ml at 20°C),ethanol,ether,90% alcohol (2000mg/ml),and water (4710mg/ml at 25°C).
SMILES [Zn+2].[Br-].[Br-]
IUPAC Name zinc;dibromide
Molecular Weight (g/mol) 225.188
Formula Weight 225.19 (Anhydrous)
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Zinc bromide hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.