missing translation for 'onlineSavingsMsg'
Learn More

Zinc bromide, Puriss., anhydrous, ≥98%, Honeywell Fluka™

Product Code. p-7071697 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
25 kg
4 x 5 kg
Unit Size:
100g
1kg
25kg
500g
5kg
Pack of 4
This item is not returnable. View return policy

Product Code. 15630570

Brand: Honeywell-Fluka 02128100G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss., anhydrous,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7699-45-8
Molecular Formula Br2Zn
Molecular Weight (g/mol) 225.188
MDL Number MFCD00011294
InChI Key VNDYJBBGRKZCSX-UHFFFAOYSA-L
Synonym zincbromide, zinc ii bromide, acmc-209p7h, zinc 2+ ion dibromide, ksc378m6n, zinc bromide hydrate, puratronic™, zinc bromide, anhydrous 100g
PubChem CID 10421208
IUPAC Name zinc;dibromide
SMILES [Zn+2].[Br-].[Br-]

Specifications

Physical Form Anhydrous
Quantity 100 g
Linear Formula ZnBr2
UN Number 3260
Formula Weight 225.2g/mol
Percent Purity ≥98%
Grade puriss.
Chemical Name or Material Zinc Bromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.